7-Oxa-2-azaspiro[3.5]nonane ≥95%

Supplier: Matrix Scientific

070662-500MG 070662-250MG 070662-100MG
101945-806EA 384.62 USD
101945-806 101945-802 101945-804
7-Oxa-2-azaspiro[3.5]nonane ≥95%
7-Oxa-2-azaspiro[3.5]nonane

Hazard: Irritant

Formula: C₇H₁₃NO
MDL Number: MFCD11204146
CAS Number: 194157-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR