6-[(tert-Butoxycarbonyl)amino]spiro[3.3]heptane-2-carboxylic acid ethyl ester ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 6-(Boc-amino)spiro[3.3]heptane-2-carboxylate
070661-500MG
070661-100MG
070661-250MG
101945-796EA
524.62
USD
101945-796
101945-798
101945-800
6-[(tert-Butoxycarbonyl)amino]spiro[3.3]heptane-2-carboxylic acid ethyl ester ≥95%
6-[(tert-Butoxycarbonyl)amino]spiro[3.3]heptane-2-carboxylic acid ethyl ester
Hazard: Irritant
Formula:
C₁₅H₂₅NO₄ MW: 283.37 g/mol |
MDL Number:
MFCD17016158 CAS Number: 1272412-66-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...