Spiro[3.3]heptane-2,6-dicarboxylicacid diethyl ester ≥95%
Supplier: Matrix Scientific
Synonyms:
Diethyl spiro[3.3]heptane-2,6-dicarboxylate
070660-1G
070660-5G
070660-500MG
101945-794EA
4343.08
USD
101945-794
101945-790
101945-792
Spiro[3.3]heptane-2,6-dicarboxylicacid diethyl ester ≥95%
Spiro[3.3]heptane-2,6-dicarboxylicacid diethyl ester
Hazard: Irritant
Formula:
C₁₃H₂₀O₄ MW: 240.3 g/mol |
MDL Number:
MFCD18432786 CAS Number: 132616-34-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...