4-Amino-1H-indazole-1-carboxylic acid tert-butyl ester ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Amino-1H-indazole, N1-BOC protectedtert-Butyl 4-amino-1H-indazole-1-carboxylate

070649-5G 070649-1G
101945-732EA 815.38 USD
101945-732 101945-734
4-Amino-1H-indazole-1-carboxylic acid tert-butyl ester ≥97%
4-Amino-1H-indazole-1-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₂H₁₅N₃O₂
MW: 233.27 g/mol
MDL Number: MFCD11052618
CAS Number: 801315-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR