Isothiazole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

070633-250MG 070633-1G
101945-666EA 623.08 USD
101945-666 101945-668
Isothiazole-3-carboxylic acid ≥95%
Isothiazole-3-carboxylic acid

Hazard: Irritant

Formula: C₄H₃NO₂S
MDL Number: MFCD09834765
CAS Number: 4576-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR