7-Oxo-2,6-diazaspiro[3.4]octane-2-carboxylic acid tert-butyl ester ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Boc-7-oxo-2,6-diaza-spiro[3.4]octane-2-carboxylate
, tert-Butyl 7-oxo-2,6-diazaspiro[3.4]octane-2-carboxylate
070627-1G
101945-642EA
330.77
USD
101945-642
7-Oxo-2,6-diazaspiro[3.4]octane-2-carboxylic acid tert-butyl ester ≥95%
7-Oxo-2,6-diazaspiro[3.4]octane-2-carboxylic acid tert-butyl ester
Hazard: Irritant
Formula:
C₁₁H₁₈N₂O₃ MW: 226.28 g/mol |
MDL Number:
MFCD18325190 CAS Number: 1234616-51-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...