6-Hydroxy-2-azaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 6-hydroxy-2-azaspiro[3.3]heptane-2-carboxylate6-Hydroxy-2-azaspiro[3.3]heptane, N-BOC protected

070624-1G
101945-626EA 746.15 USD
101945-626
6-Hydroxy-2-azaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester ≥95%
6-Hydroxy-2-azaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₁H₁₉NO₃
MW: 213.28 g/mol
MDL Number: MFCD15071431
CAS Number: 1147557-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR