2,4-Dioxo-1,3,8-triazaspiro[4.5]decane-8-carboxylic acid tert-butyl ester ≥97%

Supplier: Matrix Scientific

Synonyms: 2,4-Dioxo-1,3,8-triazaspiro[4.5]decane, N8-BOC protected8-Boc-2,4,8-triazaspiro[4.5]decane-1,3-dione

070621-1G
101945-610EA 461.54 USD
101945-610
2,4-Dioxo-1,3,8-triazaspiro[4.5]decane-8-carboxylic acid tert-butyl ester ≥97%
2,4-Dioxo-1,3,8-triazaspiro[4.5]decane-8-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₂H₁₉N₃O₄
MW: 269.3 g/mol
MDL Number: MFCD03425517
CAS Number: 183673-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR