3-Hydroxy-1-oxa-8-azaspiro[4.5]decane-8-carboxylic acid tert-butyl ester ≥97%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 3-hydroxy-1-oxa-8-azaspiro[4.5]decane-8-carboxylate
070620-500MG
070620-100MG
101945-604EA
1209.23
USD
101945-604
101945-602
3-Hydroxy-1-oxa-8-azaspiro[4.5]decane-8-carboxylic acid tert-butyl ester ≥97%
3-Hydroxy-1-oxa-8-azaspiro[4.5]decane-8-carboxylic acid tert-butyl ester
Hazard: Irritant
Formula:
C₁₃H₂₃NO₄ MW: 257.33 g/mol |
MDL Number:
MFCD09749842 CAS Number: 240401-09-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...