3-Difluoromethylpiperidine hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
3-(Difluoromethyl)piperidine hydrochloride
070613-1G
101945-572EA
1192.31
USD
101945-572
3-Difluoromethylpiperidine hydrochloride ≥95%
3-Difluoromethylpiperidine hydrochloride
Hazard: Irritant
Formula:
C₆H₁₂ClF₂N MW: 171.62 g/mol |
MDL Number:
MFCD18325187 CAS Number: 1093759-69-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...