3-Fluoro-1,3-azetidinedicarboxylic acid tert-butyl ester ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(tert-Butoxycarbonyl)-3-fluoroazetidine-3-carboxylic acid
070605-500MG
070605-250MG
101945-544EA
4246.15
USD
101945-544
101945-546
3-Fluoro-1,3-azetidinedicarboxylic acid tert-butyl ester ≥95%
3-Fluoro-1,3-azetidinedicarboxylic acid tert-butyl ester
Hazard: Irritant
Formula:
C₉H₁₄FNO₄ MW: 219.21 g/mol |
MDL Number:
MFCD15071784 CAS Number: 1126650-67-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...