1-Oxa-4,8-diazaspiro[5.5]undecane-8-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

070588-250MG 070588-500MG
101945-456EA 1629.23 USD
101945-456 101945-458
1-Oxa-4,8-diazaspiro[5.5]undecane-8-carboxylic acid tert-butyl ester ≥95%
1-Oxa-4,8-diazaspiro[5.5]undecane-8-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₃H₂₄N₂O₃
MW: 256.35 g/mol
MDL Number: MFCD11227062
CAS Number: 1160247-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR