3-Methyl-3-thietanamine ≥96%

Supplier: Matrix Scientific

070586-500MG 070586-250MG
101945-452EA 1221.54 USD
101945-452 101945-454
3-Methyl-3-thietanamine ≥96%
3-Methyl-3-thietanamine

Hazard: Irritant

Formula: C₄H₉NS
MDL Number: MFCD18325180
CAS Number: 943437-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR