2-[2,2-Difluoro-1-(MEM)ethenyl]boronic acid pinacol ester ≥94%

Supplier: Matrix Scientific

070583-1G
101945-438EA 1396.92 USD
101945-438
2-[2,2-Difluoro-1-(MEM)ethenyl]boronic acid pinacol ester ≥94%
2-[2,2-Difluoro-1-(MEM)ethenyl]boronic acid pinacol ester

Hazard: Irritant

Formula: C₁₂H₂₁BF₂O₅
MDL Number: MFCD18325179
CAS Number: 1272412-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR