1,8-Diazaspiro[5.5]undecane-1-carboxylic acid tert-butyl ester ≥95%
Supplier: Matrix Scientific
070579-500MG
070579-250MG
101945-420EA
1047.69
USD
101945-420
101945-422
1,8-Diazaspiro[5.5]undecane-1-carboxylic acid tert-butyl ester ≥95%
1,8-Diazaspiro[5.5]undecane-1-carboxylic acid tert-butyl ester
Hazard: Irritant
Formula:
C₁₄H₂₆N₂O₂ MW: 254.38 g/mol |
MDL Number:
MFCD10700119 CAS Number: 960294-18-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...