2-Oxa-9-azaspiro[5.5]undecane-9-carboxylicacid tert-butyl ester ≥95%
Supplier: Matrix Scientific
070570-500MG
070570-250MG
101945-376EA
1396.92
USD
101945-376
101945-378
2-Oxa-9-azaspiro[5.5]undecane-9-carboxylicacid tert-butyl ester ≥95%
2-Oxa-9-azaspiro[5.5]undecane-9-carboxylicacid tert-butyl ester
Hazard: Irritant
Formula:
C₁₄H₂₅NO₃ MW: 255.36 g/mol |
MDL Number:
MFCD13180582 CAS Number: 374795-47-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...