2,7-Diazaspiro[4.5]decane-7-carboxylic acid tert-butyl ester ≥96%
Supplier: Matrix Scientific
070567-250MG
070567-1G
101945-364EA
1221.54
USD
101945-364
101945-366
2,7-Diazaspiro[4.5]decane-7-carboxylic acid tert-butyl ester ≥96%
2,7-Diazaspiro[4.5]decane-7-carboxylic acid tert-butyl ester
Hazard: Irritant
Formula:
C₁₃H₂₄N₂O₂ MW: 240.35 g/mol |
MDL Number:
MFCD07779071 CAS Number: 236406-61-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...