1,7-Diazaspiro[4.4]nonane-7-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

070552-100MG 070552-500MG 070552-250MG
101945-276EA 769.23 USD
101945-276 101945-278 101945-274
1,7-Diazaspiro[4.4]nonane-7-carboxylic acid tert-butyl ester ≥95%
1,7-Diazaspiro[4.4]nonane-7-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₂H₂₂N₂O₂
MW: 226.32 g/mol
MDL Number: MFCD09266216
CAS Number: 646055-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR