4-(Tributylstannyl)-3,6-dihydro-2H-pyran ≥92%
Supplier: Matrix Scientific
070543-5G
070543-1G
101945-232EA
2692.31
USD
101945-232
101945-230
4-(Tributylstannyl)-3,6-dihydro-2H-pyran ≥92%
4-(Tributylstannyl)-3,6-dihydro-2H-pyran
Hazard: Irritant
Formula:
C₁₇H₃₄OSn |
MDL Number:
MFCD18325166 CAS Number: 535924-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...