2-Chloro-6,7-dihydro-4H-pyrano[4,3-d]thiazole ≥96%

Supplier: Matrix Scientific

070538-1G 070538-5G
101945-210EA 884.62 USD
101945-210 101945-212
2-Chloro-6,7-dihydro-4H-pyrano[4,3-d]thiazole ≥96%
2-Chloro-6,7-dihydro-4H-pyrano[4,3-d]thiazole

Hazard: Irritant

Formula: C₆H₆ClNOS
MDL Number: MFCD13189990
CAS Number: 259810-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR