3(2H)-Pyridazinone ≥97%
Supplier: Matrix Scientific
Synonyms:
2,3-dihydropyridazin-3-one
, Pyridazin-3(2H)-one
070537-5G
070537-1G
101945-208EA
320
USD
101945-208
101945-206
3(2H)-Pyridazinone ≥97%
3(2H)-Pyridazinone
Hazard: Irritant
Formula:
C₄H₄N₂O MW: 96.0874 g/mol |
MDL Number:
MFCD00099605 CAS Number: 504-30-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...