6-Chloro-3-methylimidazo[1,2-b]pyridazine ≥97%

Supplier: Matrix Scientific

070508-1G 070508-5G
101945-088EA 524.62 USD
101945-088 101945-090
6-Chloro-3-methylimidazo[1,2-b]pyridazine ≥97%
6-Chloro-3-methylimidazo[1,2-b]pyridazine

Hazard: Irritant

Formula: C₇H₆ClN₃
MDL Number: MFCD09991804
CAS Number: 137384-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR