5-Bromo-1H-benzoimidazole-2-carboxylic acid ≥96%
Supplier: Matrix Scientific
Synonyms:
5-Bromo-1H-benzo[d]imidazole-2-carboxylic acid
070501-5G
070501-1G
101945-060EA
146.15
USD
101945-060
101945-062
5-Bromo-1H-benzoimidazole-2-carboxylic acid ≥96%
5-Bromo-1H-benzoimidazole-2-carboxylic acid
Hazard: Irritant/Store in Freezer
Formula:
C₈H₅BrN₂O₂ MW: 241.05 g/mol |
MDL Number:
MFCD06738769 CAS Number: 40197-20-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...