5-Trifluoromethyl-1H-benzoimidazole-2-carboxylic acid ≥96%
Supplier: Matrix Scientific
070500-5G
070500-1G
101945-056EA
653.85
USD
101945-056
101945-058
5-Trifluoromethyl-1H-benzoimidazole-2-carboxylic acid ≥96%
5-Trifluoromethyl-1H-benzoimidazole-2-carboxylic acid
Hazard: Irritant/Refrigerate
Formula:
C₉H₅F₃N₂O₂ MW: 230.15 g/mol |
MDL Number:
MFCD11052827 CAS Number: 2107-39-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...