4,4-Dimethoxytetrahydro-4H-pyran

Supplier: Matrix Scientific

Synonyms: 4,4-dimethoxyoxane

070490-1G 070490-5G
101945-016EA 349.23 USD
101945-016 101945-018
4,4-Dimethoxytetrahydro-4H-pyran
4,4-Dimethoxytetrahydro-4H-pyran

Hazard: Irritant

Formula: C₇H₁₄O₃
MW: 146.186 g/mol
MDL Number: MFCD00040530
CAS Number: 28218-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR