4-Fluoro-1H-indazole ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Fluoroindazole
070479-5G
070479-1G
101944-972EA
100
USD
101944-972
101944-974
4-Fluoro-1H-indazole ≥97%
4-Fluoro-1H-indazole
Hazard: Irritant
Formula:
C₇H₅FN₂ MW: 136.1264 g/mol |
MDL Number:
MFCD07371561 CAS Number: 341-23-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...