6-Benzyl-5,6,7,8-tetrahydro-1,6-naphthyridin-2(1H)-one ≥97%
Supplier: Matrix Scientific
070474-5G
070474-1G
101944-958EA
7909.23
USD
101944-958
101944-956
6-Benzyl-5,6,7,8-tetrahydro-1,6-naphthyridin-2(1H)-one ≥97%
6-Benzyl-5,6,7,8-tetrahydro-1,6-naphthyridin-2(1H)-one
Hazard: Irritant
Formula:
C₁₅H₁₆N₂O |
MDL Number:
MFCD11518944 CAS Number: 601514-58-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...