Thiomorpholine-1-oxide hydrochloride ≥95%
Supplier: Matrix Scientific
070469-5G
070469-1G
101944-936EA
3515.38
USD
101944-936
101944-934
Thiomorpholine-1-oxide hydrochloride ≥95%
Thiomorpholine-1-oxide hydrochloride
Hazard: Irritant
Formula:
C₄H₁₀ClNOS |
MDL Number:
MFCD09026928 CAS Number: 76176-87-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...