tert-Butyl-2-chloro-7,8-dihydropyrido[4,3-d]pyrimidine-6(5H)-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 2-chloro-7,8-dihydropyrido[4,3-d]pyrimidine-6(5H)-carboxylate

070468-1G
101944-932EA 969.23 USD
101944-932
tert-Butyl-2-chloro-7,8-dihydropyrido[4,3-d]pyrimidine-6(5H)-carboxylate ≥95%
tert-Butyl-2-chloro-7,8-dihydropyrido[4,3-d]pyrimidine-6(5H)-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₆ClN₃O₂
MW: 269.73 g/mol
MDL Number: MFCD11518980
CAS Number: 1092352-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR