5-Bromo-2,4-dimethyl-2H-indazole

Supplier: Matrix Scientific

070467-1G 070467-5G
101944-928EA 1978.46 USD
101944-928 101944-930
5-Bromo-2,4-dimethyl-2H-indazole
5-Bromo-2,4-dimethyl-2H-indazole

Hazard: Irritant

Formula: C₉H₉BrN₂
MDL Number: MFCD12028635
CAS Number: 1159511-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR