2-(Methylthio)-5-(tributylstannyl)thiazole ≥96%

Supplier: Matrix Scientific

070459-500MG 070459-5G 070459-1G
101944-894EA 7909.23 USD
101944-894 101944-892 101944-890
2-(Methylthio)-5-(tributylstannyl)thiazole ≥96%
2-(Methylthio)-5-(tributylstannyl)thiazole

Hazard: Irritant

Formula: C₁₆H₃₁NS₂Sn
MDL Number: MFCD16140398
CAS Number: 157025-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR