2-(5-(Tributylstannyl)thiazol-2-yl)-2-propanol ≥95%
Supplier: Matrix Scientific
070456-500MG
070456-1G
070456-5G
101944-874EA
1538.46
USD
101944-874
101944-872
101944-876
2-(5-(Tributylstannyl)thiazol-2-yl)-2-propanol ≥95%
2-(5-(Tributylstannyl)thiazol-2-yl)-2-propanol
Hazard: Irritant
Formula:
C₁₈H₃₅NOSSn |
MDL Number:
MFCD16140395 CAS Number: 1245816-17-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...