(2S,4S)-4-Fluoropyrrolidine-2-carboxamide hydrochloride ≥97%
Supplier: Matrix Scientific
Synonyms:
cis-4-Fluoro-L-prolinamide hydrochloride
070444-5G
070444-1G
070444-25G
101944-804EA
6410.77
USD
101944-804
101944-802
101944-800
(2S,4S)-4-Fluoropyrrolidine-2-carboxamide hydrochloride ≥97%
(2S,4S)-4-Fluoropyrrolidine-2-carboxamide hydrochloride
Hazard: Irritant
Formula:
C₅H₁₀ClFN₂O MW: 168.6 g/mol |
MDL Number:
MFCD07368567 CAS Number: 426844-23-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...