2-Iodo-5-methoxyaniline ≥95%

Supplier: Matrix Scientific

070427-5G
101944-706EA 438.46 USD
101944-706
2-Iodo-5-methoxyaniline ≥95%
2-Iodo-5-methoxyaniline

Hazard: Irritant

Formula: C₇H₈INO
MW: 249.0490 g/mol
Melting Pt: 33…36 °C
MDL Number: MFCD08458361
CAS Number: 153898-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR