4-Azidobutyric acid ≥95%

Supplier: Matrix Scientific

069400-1G 069400-500MG
101939-952EA 1044.62 USD
101939-952 101939-950
4-Azidobutyric acid ≥95%
4-Azidobutyric acid

Hazard: Irritant/Refrigerate

Formula: C₄H₇N₃O₂
MDL Number: MFCD11850695
CAS Number: 54447-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR