(1-Benzyl-3-fluoro-3-piperidyl)methanamine ≥90%

Supplier: Matrix Scientific

069386-500MG 069386-250MG
101939-878EA 1629.23 USD
101939-878 101939-880
(1-Benzyl-3-fluoro-3-piperidyl)methanamine ≥90%
(1-Benzyl-3-fluoro-3-piperidyl)methanamine

Hazard: Irritant

Formula: C₁₃H₁₉FN₂
MDL Number: MFCD15071766
CAS Number: 1185749-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR