Ethyl-3-oxocyclobutanecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 3-oxocyclobutanecarboxylate
069356-25G
069356-5G
069356-1G
101939-758EA
60
USD
101939-758
101939-756
101939-760
Ethyl-3-oxocyclobutanecarboxylate ≥95%
Ethyl-3-oxocyclobutanecarboxylate
Hazard: Irritant
Formula:
C₇H₁₀O₃ MW: 142.154 g/mol |
MDL Number:
MFCD09834114 CAS Number: 87121-89-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...