2,4-Piperidinedicarboxylic acid ≥95%

Supplier: Matrix Scientific

069349-250MG
101939-728EA 215.38 USD
101939-728
2,4-Piperidinedicarboxylic acid ≥95%
2,4-Piperidinedicarboxylic acid

Hazard: Irritant

Formula: C₇H₁₁NO₄
MDL Number: MFCD12026391
CAS Number: 85819-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR