(3-Methylthietan-3-yl)methanol ≥95%

Supplier: Matrix Scientific

069344-250MG 069344-1G 069344-500MG
101939-718EA 769.23 USD
101939-718 101939-716 101939-714
(3-Methylthietan-3-yl)methanol ≥95%
(3-Methylthietan-3-yl)methanol

Hazard: Irritant

Formula: C₅H₁₀OS
MDL Number: MFCD19982771
CAS Number: 57523-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR