Ethyl-1-benzyl-4-bromo-3-oxopiperidine-4-carboxylate hydrobromide ≥95%

Supplier: Matrix Scientific

069341-500MG 069341-250MG
101939-708EA 1098.46 USD
101939-708 101939-706
Ethyl-1-benzyl-4-bromo-3-oxopiperidine-4-carboxylate hydrobromide ≥95%
Ethyl-1-benzyl-4-bromo-3-oxopiperidine-4-carboxylate hydrobromide

Hazard: Irritant

Formula: C₁₅H₁₉Br₂NO₃
MDL Number: MFCD18909955
CAS Number: 1303972-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR