Ethyl-5,5,5-trifluoro-4-oxopentanoate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5,5,5-trifluoro-4-oxopentanoate

069334-250MG 069334-500MG
101939-684EA 1073.85 USD
101939-684 101939-682
Ethyl-5,5,5-trifluoro-4-oxopentanoate ≥95%
Ethyl-5,5,5-trifluoro-4-oxopentanoate

Hazard: Irritant

Formula: C₇H₉F₃O₃
MW: 198.14 g/mol
MDL Number: MFCD12828184
CAS Number: 70961-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR