1-Adamantaneethylsulfonamide ≥95%

Supplier: Matrix Scientific

069329-250MG 069329-500MG
101939-662EA 2472.31 USD
101939-662 101939-660
1-Adamantaneethylsulfonamide ≥95%
1-Adamantaneethylsulfonamide

Hazard: Irritant

Formula: C₁₂H₂₁NO₂S
MDL Number: MFCD18800651
CAS Number: 1283719-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR