2-Chloro-N-(2-chloroethyl)-N-(cyclopropylmethyl)ethanamine hydrochloride ≥95%
Supplier: Matrix Scientific
069327-250MG
069327-500MG
101939-658EA
798.46
USD
101939-658
101939-656
2-Chloro-N-(2-chloroethyl)-N-(cyclopropylmethyl)ethanamine hydrochloride ≥95%
2-Chloro-N-(2-chloroethyl)-N-(cyclopropylmethyl)ethanamine hydrochloride
Hazard: Irritant
Formula:
C₈H₁₆Cl₃N |
MDL Number:
MFCD18909746 CAS Number: 90202-51-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...