3-Hydroxy-4,5,6,7-tetrahydro-2H-pyrazolo[3,4-c]pyridine-5-carboxylic acid hydrochloride ≥95%

Supplier: Matrix Scientific

069315-500MG 069315-250MG
101939-624EA 4369.23 USD
101939-624 101939-622
3-Hydroxy-4,5,6,7-tetrahydro-2H-pyrazolo[3,4-c]pyridine-5-carboxylic acid hydrochloride ≥95%
3-Hydroxy-4,5,6,7-tetrahydro-2H-pyrazolo[3,4-c]pyridine-5-carboxylic acid hydrochloride

Hazard: Irritant

Formula: C₇H₁₀ClN₃O₃
MDL Number: MFCD19443957
CAS Number: 1313739-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR