4-Fluoromethylpiperidine hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
4-(Fluoromethyl)piperidine hydrochloride
069305-250MG
069305-500MG
101939-596EA
700
USD
101939-596
101939-594
4-Fluoromethylpiperidine hydrochloride ≥95%
4-Fluoromethylpiperidine hydrochloride
Hazard: Irritant
Formula:
C₆H₁₃ClFN MW: 153.63 g/mol |
MDL Number:
MFCD19443952 CAS Number: 787564-27-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...