4-Chloro-5-methylpyrrolo[2,1-f][1,2,4]triazine ≥95%

Supplier: Matrix Scientific

069278-1G 069278-500MG
101939-568EA 3570.77 USD
101939-568 101939-566
4-Chloro-5-methylpyrrolo[2,1-f][1,2,4]triazine ≥95%
4-Chloro-5-methylpyrrolo[2,1-f][1,2,4]triazine

Hazard: Irritant

Formula: C₇H₆ClN₃
MDL Number: MFCD12924549
CAS Number: 529508-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR