3-(4-Ethylphenyl)tetrahydrofuran ≥95%

Supplier: Matrix Scientific

069269-5G 069269-1G
101939-538EA 3076.92 USD
101939-538 101939-536
3-(4-Ethylphenyl)tetrahydrofuran ≥95%
3-(4-Ethylphenyl)tetrahydrofuran

Hazard: Irritant

Formula: C₁₂H₁₆O
MDL Number: MFCD19443947
CAS Number: 100058-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR