3-(Chloromethyl)-3-(iodomethyl)oxetane ≥95%
Supplier: Matrix Scientific
069262-500MG
069262-250MG
101939-512EA
1623.08
USD
101939-512
101939-510
3-(Chloromethyl)-3-(iodomethyl)oxetane ≥95%
3-(Chloromethyl)-3-(iodomethyl)oxetane
Hazard: Irritant
Formula:
C₅H₈ClIO |
MDL Number:
MFCD01755639 CAS Number: 35842-61-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...