3-Fluoro-3-nitrooxetane ≥95%

Supplier: Matrix Scientific

069259-500MG 069259-250MG
101939-504EA 2252.31 USD
101939-504 101939-502
3-Fluoro-3-nitrooxetane ≥95%
3-Fluoro-3-nitrooxetane

Hazard: Irritant

Formula: C₃H₄FNO₃
MDL Number: MFCD17170026
CAS Number: 70187-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR