Isoquinoline-4-carbaldehyde ≥97%
Supplier: Matrix Scientific
Synonyms:
Isoquinoline-4-carboxaldehyde
069247-5G
069247-1G
101939-460EA
718.75
USD
101939-460
101939-458
Isoquinoline-4-carbaldehyde ≥97%
Isoquinoline-4-carbaldehyde
Hazard: Irritant
Formula:
C₁₀H₇NO MW: 157.17 g/mol Melting Pt: 101…106 °C |
MDL Number:
MFCD00829440 CAS Number: 22960-16-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...